Natriumfluorid-Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

919-25ML
919-25MLEA 107 EUR
919-25ML
Natriumfluorid-Lösung, Sigma-Aldrich®
Sodium fluoride

For use in inhibiting α-naphthyl acetate reaction on monocytes. Optional reagent in Sigma Esterase Procedure 91A.

Formel: NaF
Molecular Weight: 41,99 g/mol
Dichte: 1,005 g/mL (25 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003524
CAS-Nummer: 7681-49-4
IVDLogo

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR