Biocytin TMR, CF® 488A Dye

Lieferant: Biotium

Synonyme: 5-(&-6)-Tetramethylrhodamine Biocytin, Biocytin conjugates

92004
92004.EA 535 EUR
92004.
Biocytin TMR, CF® 488A Dye
Biocytin TMR

CF®488A is a superior green fluorescent dye


  • Minimally charged, for less non-specific binding than Alexa Fluor® 488
  • Narrower emission spectrum for less bleed to red
  • Very photostable
  • Compatible with super-resolution imaging by TIRF
  • Highly water soluble and pH-insensitive


CF® dye biocytin is a cell-impermeant fixable polar tracer that is a conjugate of CF® dye and biotin with an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis.


Stock solutions may be prepared in DMF or DMSO.


CF® dye biocytin is available in the following colors: CF®488A, CF®568, CF®594, and CF®640R.


Bitte beachten Store at –20 °C, protected from light.

Bestellen

Spezifikation Testergebnisse

Abs/Em Maxima 490/515 nm
Extinction coefficient 70,000
Molecular weight ~914
Excitation laser line 488 nm
Replaces Alexa Fluor® 488, DyLight® 488, fluorescein (aka FITC, FAM), Cy®2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR