Biocytin TMR, CF® 568 Dye

Lieferant: Biotium

Synonyme: 5-(&-6)-Tetramethylrhodamine Biocytin, Biocytin conjugates

92005
92005.EA 535 EUR
92005.
Biocytin TMR, CF® 568 Dye
Biocytin TMR

CF®568, an Outshines Alexa Fluor®568


  • Yields much brighter antibody conjugates than Alexa Fluor® 568
  • Extremely photostable
  • Excellent choice for multi-color imaging with CF®488A and CF®640R
  • Compatible with TIRF and multicolor STORM superresolution imaging


CF® dye biocytin is a cell-impermeant fixable polar tracer that is a conjugate of CF® dye and biotin with an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis.


Stock solutions may be prepared in DMF or DMSO.


CF® dye biocytin is available in the following colors: CF®488A, CF®568, CF®594, and CF®640R.


Bitte beachten Store at –20 °C, protected from light.

Bestellen

Spezifikation Testergebnisse

Abs/Em Maxima 562/583 nm
Extinction coefficient 100,000
Molecular weight ~ 714
Excitation laser line 532 nm or 568 nm
Direct replacement for Alexa Fluor® 568, ATTO 565, Rhodamine Red

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR