Biocytin TMR, CF® 594 Dye

Lieferant: Biotium

Synonyme: 5-(&-6)-Tetramethylrhodamine Biocytin, Biocytin conjugates

92006
92006.EA 535 EUR
92006.
Biocytin TMR, CF® 594 Dye
Biocytin TMR

CF®594, Truly the brightest deep red dye


  • Yields the brightest antibody conjugates among spectrally similar dyes.
  • Excellent choice for multicolor imaging with green dyes like CF®488A
  • Extremely photostable
  • Also see CF®594ST, a version of CF®594 engineered specifically for STORM microcopy


CF® dye biocytin is a cell-impermeant fixable polar tracer that is a conjugate of CF® dye and biotin with an aldehyde-fixable primary amine. Polar tracers are commonly used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis.


Stock solutions may be prepared in DMF or DMSO.


CF® dye biocytin is available in the following colors: CF®488A, CF®568, CF®594, and CF®640R.


Bitte beachten Store at –20 °C, protected from light.

Bestellen

Spezifikation Testergebnisse

Abs/Em Maxima 593/614 nm
Extinction coefficient 115,000
Molecular weight ~730
Excitation laser line 532 nm, 568 nm or 594 nm
Replaces Alexa Fluor® 594, DyLight® 594, Texas Red®

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR