Photoreactive dyes and reagents
Lieferant: Biotium
92016
92016.EA 304 EUR
92016.
Photoreactive dyes and reagents
Proteinkonjugate und Markierungen
INA (5-iodonaphthyl-1-azide) is a lipophilic photoreactive probe, which has been used to selectively label membrane-embedded cysteine residues of proteins.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbea1a6ec66b0343583b8ccc0fb1b7bc10e196cb&vid=e4b1e5885ff40ea4362872a51294c0e3728a8a40&prid=web&clid=SAPDAM