Periodic table mix 1 for ICP, 33 elements, TraceCERT®, Supelco®
Lieferant: Merck
92091-100ML
92091-100MLEA 275 EUR
92091-100ML
Periodic table mix 1 for ICP, 33 elements, TraceCERT®, Supelco®
Standards Kationischer Standards Mehrelement-Standards
Periodic table mix 1 for ICP used as a standard for the quantitative determination of mineral composition in Tenualosa ilisha using inductively coupled plasma atomic emission spectroscopy (ICP-AES).
It contains Al, As, Ba, Be, Bi, B, Ca, Cd, Cs, Cr, Co, Cu, Ga, In, Fe, Pb, Li, Mg, Mn, Ni, P, K, Rb, Se, Si, Ag, Na, Sr, S, Te, Tl, V and Zn in 10% nitric acid (contains HF traces).
Standards: Certified in accordance with ISO/IEC 17025 and ISO 17034