Cyclohexanon ≥99,0% (durch GC), BAKER ANALYZED®, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Pimelic Ketone
9210-03EA
283.81
EUR
9210-03
Cyclohexanon ≥99,0% (durch GC), BAKER ANALYZED®, J.T.Baker®
Cyclohexanone
Lot analysis on label.
Formel:
C₆H₁₀(=O) Molecular Weight: 98,14 g/mol Siedepunkt: 155,6 °C (1013 hPa) Schmelzpunkt: –31 °C Dichte: 0,9478 g/cm³ (20 °C) Flammpunkt: 43 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001625 CAS-Nummer: 108-94-1 EINECS: 203-631-1 UN: 1915 ADR: 3,III REACH: 01-2119453616-35 Merck Index: 13,02755 |
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use |
|
Assay (C₆H₁₀O) (by GC) | ≥ 99.0 % |
Color (APHA) | ≤ 10 |
Density (g/mL) at 25°C | 0.940 - 0.943 |
Residue after Evaporation | ≤ 0.0500 % |
Acidity (meq/g) | ≤ 0.003 |
Water (H₂O)(by Karl Fischer titrn) | ≤ 0.1 % |
Identification (by IR) | Passes Test |
NOTE: Color reported is at time of packaging; material will darken with age. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...