Kieselgel, TECHNICAL Drying_agent, with moisture indicator_blue gel
Lieferant: VWR Chemicals
|
Gefahr
|
92120.0120EA
0
EUR
92120.0120
92120.9012
Kieselgel, TECHNICAL Drying_agent, with moisture indicator_blue gel
Silica gel
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 Schmelzpunkt: 1710 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 63231-67-4 |
Spezifikation Testergebnisse
Appearance | Blue granules |
Loss on drying (145°C) | Max. 2 % |
Absorption capacity (100% RH) | Min. 30 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...