Biotin SE protein labelling kit
Lieferant: Biotium
92224
92224.EA
707
EUR
92224.
Biotin SE protein labelling kit
Proteinkonjugate und Markierungen
This kit contains everything needed to label antibodies or other proteins with biotin succinimidyl ester (biotin SE). Each kit contains reagents for three labelling reactions of up to 1 mg protein per reaction.