Antibody labelling kits, Mix-n-Stain™ CF™ Dye
Lieferant: Biotium
92230 92231 92232 92233 92234 92235 92236 92237 92238 92239 92240 92241 92242 92243 92245 92246 92253 92258 92260 92261 92262 92263 92250 92251 92252 92254 92255 92256 92257 92259 92264 92265 92270 92271 92272 92273 92274 92275 92276 92277 92278 92279 92280 92281 92282 92283 92284 92285 92290 92287 92267 92247 92291 92289 92248 92268 92288 92303 92304 92305 92331 92332 92333 92316 92317 92318 92319 92320 92321 92322 92323 92324 92334 92335 92336 92337 92338 92339 92425 92426 92427 92428 92429 92430 92431 92432 92433
92230.EA 224 EUR
92230. 92231. 92232. 92233. 92234. 92235. 92236. 92237. 92238. 92239. 92240. 92241. 92242. 92243. 92245. 92246. 92253. 92258. 92260. 92261. 92262. 92263. 92250. 92251. 92252. 92254. 92255. 92256. 92257. 92259. 92264. 92265. 92270. 92271. 92272. 92273. 92274. 92275. 92276. 92277. 92278. 92279. 92280. 92281. 92282. 92283. 92284. 92285. 92290. 92287. 92267. 92247. 92291. 92289. 92248. 92268. 92288. 92303. 92304. 92305. 92331. 92332. 92333. 92316. 92317. 92318. 92319. 92320. 92321. 92322. 92323. 92324. 92334. 92335. 92336. 92337. 92338. 92339. 92425. 92426. 92427. 92428. 92429. 92430. 92431. 92432. 92433.
Antibody labelling kits, Mix-n-Stain™ CF™ Dye
Proteinkonjugate und Markierungen
Label your antibody with one of our superior CF® Dyes in 30 minutes without a purification step. Labelling tolerates many common buffer components including BSA and ascites.
  • Label 5 to 20 µg, 20 to 50 µg, or 50 to 100 µg Ab
  • Choice of 29 CF® Dye colours
  • Less than 30 seconds of hands-on time
  • Total reaction time of 30 minutes
  • No purification, 100% recovery
  • Compatible with BSA, gelatin, ascites
Mix-n-Stain™ CF® Dye antibody labelling kits allow you to label ≤5 µg up to 100 µg of your antibody in just 30 minutes, with minimal hands-on time and no purification.
Mix-n-Stain™ labeling tolerates common antibody buffer formulations. An ultrafiltration centrifuge vial is provided to quickly remove interfering substances like glycerol, or to concentrate your antibody. With a slight modification in protocol, antibodies can be labelled in the presence of BSA, gelatin, or ascites fluid. Because CF® Dyes are highly water soluble, the presence of other proteins like BSA or gelatin in the labelling reaction has minimal effect on background fluorescence, because any labelled non-antibody proteins readily wash away during immunofluorescence staining.
These kits feature a choice of 32 of Biotium’s next-generation CF® Dyes, which were designed for advantages in brightness and photostability compared to Alexa Fluor®, DyLight®, and other fluorescent dyes.
Mix-n-Stain™ antibody labelling kits are very simple to use (see the workflow overview). But before you begin, you must check that your antibody meets the compatibility requirements for labelling, and choose the right labelling protocol. See the product table below for a quick reference of the maximum excitation and emission of each CF® Dye.
Biotium now offer Mix-n-Stain™ Maxi Antibody labelling kits for labelling 1 mg of antibody with CF® Dyes, and Mix-n-Stain™ Nanobody labelling kits. Mix-n-Stain™ kits also are available for labelling antibodies with enzymes (HRP, AP, and GOx), fluorescent proteins (PE, APC, PerCP, and tandem dyes), biotin, or DIG.
Lieferumfang: Kit includes ultrafiltration spin vial (for antibody concentration or buffer exchange, if needed), 10X Mix-n-Stain™ reaction buffer, lyophilised reactive CF® Dye and Mix-n-Stain™ storage buffer.