Chloride ionophore IV, Selectophore™ Ionophor, Supelco®

Lieferant: Merck

Synonyme: 4,5-Bis-[N′-(butyl)thioureido]-2,7-di-tert-butyl-9,9-dimethylxanthene

M-Clarity™ Qualitätsstufe = MQ100

92332-25MG-F
92332-25MG-FEA 545 EUR
92332-25MG-F
Chloride ionophore IV, Selectophore™ Ionophor, Supelco®
Chloride ionophore IV
Formel: C₃₃H₅₀N₄OS₂
Molecular Weight: 582.91 g/mol
MDL: MFCD06201021
CAS-Nummer: 187404-67-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c99958e5661e410266600b984715f6399e66e1a&vid=453e7aced73e1a492b93f6a4b5342fd794658e72&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR