Supelco® TraceCERT® Organic Reference Standard, ᴅʟ-α-Tocopherol acetate
Lieferant: Merck
92359-50MG
92359-50MGEA 65.2 EUR
92359-50MG
Supelco® TraceCERT® Organic Reference Standard, ᴅʟ-α-Tocopherol acetate
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.