Acetonitril ≥99,9%, HiPerSolv CHROMANORM®, PFAS-Qualität für die LC-MS
Lieferant: VWR Chemicals
|
Gefahr
|
Synonyme:
Ethannitril, Essigsäurenitril, Ethansäurenitril, Methylcyanid
Unter Stickstoff abgepackt
92497.320EA
418
EUR
92497.320
Acetonitril ≥99,9%, HiPerSolv CHROMANORM®, PFAS-Qualität für die LC-MS
Acetonitril
VWR® PFAS grade LC-MS solvents undergo a rigorous use test to ensure suitability for analysis of poly- and perfluoroalkyl substances resulting in an ultra low background of PFAS analytes. Using complementary methods a broad range of PFAS analytes are quantified to their detection limits by LC-MS.
- Specifically tested for PFAS analysis
- Ultra low background solvent for LC-MS analysis of PFAS (poly- and perfluoroalkyl substances)
Formel:
H₃CCN Molecular Weight: 41,05 g/mol Siedepunkt: 81,6 °C (1013 hPa) Schmelzpunkt: –45,7 °C Dichte: 0,782 g/cm³ (20 °C) Flammpunkt: 5 °C (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001878 CAS-Nummer: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II REACH: 01-2119471307-38 Merck Index: 13,00071 |
Spezifikation Testergebnisse
Gehalt (auf wasserfreier Substanz) | Min. 99,9 % |
Sauer reagierende Substanzen | Max. 0,0005 meq/g |
Verdampfungsrückstand | Max. 5 ppm |
Wasser | Max. 0,02 % |
Alkalinity [meq/g] | max. 0.00006 |
Gradient use test (205 nm) | max. 0.002 AU |
Gradient use test (254 nm) | max. 0.0005 AU |
Baseline drift (205 nm) | max. 0.02 AU |
Fluorescence (as quinine; 254 nm) | max. 1 ppb |
Fluorescence (as quinine; 365 nm) | max. 1 ppb |
UV transmission (190 nm) | min. 18% |
UV transmission (193 nm) | min. 50% |
UV transmission (197 nm) | min. 80% |
UV transmission (215 nm) | min. 95% |
UV transmission (230-400 nm) | min. 99% |
Ag (Silver) | max. 10 ppb |
Al (Aluminium) | max. 25 ppb |
Ca (Calcium) | max. 25 ppb |
Cu (Copper) | max. 10 ppb |
Fe (Iron) | max. 10 ppb |
K (Potassium) | max. 10 ppb |
Mg (Magnesium) | max. 10 ppb |
Mn (Manganese) | max. 10 ppb |
Na (Sodium) | max. 50 ppb |
Ni (Nickel) | max. 10 ppb |
Pb (Lead) | max. 10 ppb |
Zn (Zinc) | max. 10 ppb |
Suitability for PFAS analysis | passes test |
Suitability for LC-MS | passes test |
manufactured and filled under nitrogen |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...