Ammoniumacetat 5mM in Wasser, HiPerSolv CHROMANORM®, PFAS-Qualität Eluens, für die LC-MS

Lieferant: VWR Chemicals

Synonyme: Essigsäure Ammoniumsalz

92500.290EA 238 EUR
92500.290
Ammoniumacetat 5mM in Wasser, HiPerSolv CHROMANORM®, PFAS-Qualität Eluens, für die LC-MS
Ammoniumacetat

VWR® PFAS grade LC-MS solvents undergo a rigorous use test to ensure suitability for analysis of poly- and perfluoroalkyl substances resulting in an ultra low background of PFAS analytes. Using complementary methods a broad range of PFAS analytes are quantified to their detection limits by LC-MS.


  • Specifically tested for PFAS analysis
  • Ultra low background solvent for LC-MS analysis of PFAS (poly- and perfluoroalkyl substances)

Formel: H₃CCOONH₄
Molecular Weight: 77,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00013066
CAS-Nummer: 631-61-8
EINECS: 231-791-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Gehalt (auf wasserfreier Substanz) Min. 99,9 %
Sauer reagierende Substanzen Max. 0,0005 meq/g
Verdampfungsrückstand Max. 5 ppm
Wasser Max. 0,02 %
Na (Sodium) max. 0.5 ppm
Suitability for PFAS analysis passes test
manufactured and filled under nitrogen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR