Aceton ≥99,4% (durch GC, auf Wasserinhalt korrigiert), ULTRA RESI-ANALYZED® für die Analyse von organischen Rückständen, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

Verpackt unter Schutzgas

9254.1000EA 124 EUR
9254.1000 9254.2500 9254-03
Aceton ≥99,4% (durch GC, auf Wasserinhalt korrigiert), ULTRA RESI-ANALYZED® für die Analyse von organischen Rückständen, J.T.Baker®
Aceton

BAKER ULTRA RESI-ANALYZED® Reagent—ultra high-purity solvents and salts developed for organic residue extraction/concentration procedures.


These solvents are fully characterised and lot controlled by ECD, FID, or other method-specific detectors. Advanced stabiliser systems are incorporated to ensure solvent stability and superior performance. Solvents are suitable for exacting EPA sample cleanup protocols.

Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use Min. 99.4 %
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.4 %
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Substances Reducing Permanganate Passes Test
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.6
Water (H₂O) ≤0.5 %
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤10
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR