Supelco® TraceCERT® Organic Reference Standard, mono-Methyl fumarate
Lieferant: Merck
92571-50MG
92571-50MGEA
173
EUR
92571-50MG
Supelco® TraceCERT® Organic Reference Standard, mono-Methyl fumarate
Standards
Organische Standards
- GHS signal: GHS05
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P264 - P280 - P305 + P351 + P338 - P337 + P313