Supelco® TraceCERT® Organic Reference Standard, 4-Hydroxybenzoic acid
Lieferant: Merck
92596-50MG
92596-50MGEA
85
EUR
92596-50MG
Supelco® TraceCERT® Organic Reference Standard, 4-Hydroxybenzoic acid
Standards
Organische Standards
- GHS signal: GHS05, GHS07
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P280 - P305 + P351 + P338 + P310