5,5-Dimethyl-1-pyrroline-N-oxide ≥98,0% (durch GC) für ESR-Spektroskopie, Supelco®
Lieferant: Merck
Synonyme:
DMPO, 5,5-Dimethyl-1-pyrroline N-oxide
92688-100MG
92688-100MGEA
117
EUR
92688-100MG
5,5-Dimethyl-1-pyrroline-N-oxide ≥98,0% (durch GC) für ESR-Spektroskopie, Supelco®
5,5-Dimethyl-1-pyrroline-N-oxide
5,5-Dimethyl-1-pyrroline N-oxide is a reagent generally used either as a free-radical spin-trapping agent, or electrophilic component during the synthesis of pyrrolidine derivatives. It may also be considered as 1,3-dipole in cycloaddition processes.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
Formel:
C₆H₁₁NO Molecular Weight: 113,16 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005279 CAS-Nummer: 3317-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...