Glove bags, large
Lieferant: Thermo Fisher Scientific
92717.LK 92717.LL 92727.LK 92727.LL 93727.LK 93727.LL 93737.LK 93737.LL
92717.LKEA 138 EUR
92717.LK 92717.LL 92727.LK 92727.LL 93727.LK 93727.LL 93737.LK 93737.LL
Glove bags, large
Handschuhboxen und -beutel
These polyethylene glove bags are used for handling air and moisture sensitive chemicals in laboratories. Also suitable for field sampling.
  • Minimise work exposure
  • Preserve the integrity of the sample during storage
  • Serve as a portable laboratory for conducting tests and crime scene investigations
Lieferumfang: Supplied with closure, tubing coupler, and cotton gloves.