Trimethylsulphonium hydroxide 0.25 M in Methanol, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
92732-10X1ML
92732-10X1MLEA
290
EUR
92732-10X1ML
Trimethylsulphonium hydroxide 0.25 M in Methanol, Supelco®
Trimethylsulphonium Hydroxide
Reagent for the methylation of nucleophiles such as carboxylic acids, alcohols, thiols, N-heterocycles. Rapid derivatisation of acids for GC by pre-column transesterification of glycerides.
- Ideal for GC derivatisation
Formel:
C₃H₁₀OS Flammpunkt: 11 °C |
MDL:
MFCD00216756 CAS-Nummer: 17287-03-5 |
|
Spezifikation Testergebnisse
Concentration | ~0,25 M in methanol |
Storage temperature | 2 - 8 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...