Ethylacetat ≥99,9% (durch GC, auf Wasserinhalt korrigiert), BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

9280-REA 0 EUR
9280-R
Ethylacetat ≥99,9% (durch GC, auf Wasserinhalt korrigiert), BAKER ANALYZED® ACS, J.T.Baker®
Ethylacetat

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formel: CH₃COOC₂H₅
Siedepunkt: 77,1 °C (1013 hPa)
Schmelzpunkt: –83 °C
Dichte: 0,902 g/cm³ (20 °C)
Flammpunkt: –4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.9 %
Assay (CH₃COOC₂H₅) (by GC, corrected for water) ≥99.9 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Titrable Acid (µeq/g) ≤0.9
Water (by KF, volumetric) ≤0.06 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR