9-Aminoacridin ≥99,5% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®
Lieferant: Merck
Synonyme:
9-AA
92817-1G
92817-1GEA
79.8
EUR
92817-1G
9-Aminoacridin ≥99,5% (durch HPLC) Matrixsubstanz für MALDI-MS, Supelco®
9-Aminoacridine
9-Aminoacridine, an aminoacridine dye which acts as a potent mutagen in viruses and bacteria.
A MALDI matrix exhibiting low background and good sensitivity in the analysis of low molecular weight anions, such as metabolites, in negative mode.
Formel:
C₁₃H₁₀N₂ Molecular Weight: 194,24 g/mol Schmelzpunkt: 238…240 °C |
MDL:
MFCD00037839 CAS-Nummer: 90-45-9 |
Spezifikation Testergebnisse
Appearance (Color) | Off White to Yellow |
Appearance (Form) | Powder or Crystals |
Purity (HPLC area %) | ≥ 99.5 % |
Melting Point | 238 - 240 C |
Loss on Drying | ≤ 10 % |
SULFATED ASH | ≤ 0.05 % |
Proton NMR Spectrum | Conforms to structure |
Metal trace Analysis (ICP) | Corresponds to requirements |
BARIUM (ICP) | ≤ 5.0 mg/kg |
CALCIUM (ICP) | ≤ 200 mg/kg |
CADMIUM (ICP) | ≤ 5.0 mg/kg |
COBALT (ICP) | ≤ 5.0 mg/kg |
CHROMIUM (ICP) | ≤ 5.0 mg/kg |
COPPER (ICP) | ≤ 5.0 mg/kg |
IRON (ICP) | ≤ 10 mg/kg |
POTASSIUM (ICP) | ≤ 50 mg/kg |
LITHIUM (ICP) | ≤ 5.0 mg/kg |
MAGNESIUM (ICP) | ≤ 5.0 mg/kg |
MANGANESE (ICP) | ≤ 5.0 mg/kg |
SODIUM (ICP) | ≤ 50 mg/kg |
NICKEL (ICP) | ≤ 5.0 mg/kg |
LEAD (ICP) | ≤ 5.0 mg/kg |
ZINC (ICP) | ≤ 5.0 mg/kg |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...