Ethylenglycol ≥99,0% (durch GC), BAKER ANALYZED®, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Glycol, 1,2-Dihydroxyethan, 1,2-Ethandiol
9300-03EA
320.46
EUR
9300-03
9300-01
9300-07
9300-R
Ethylenglycol ≥99,0% (durch GC), BAKER ANALYZED®, J.T.Baker®
Ethylenglycol
Formel:
HOCH₂CH₂OH Siedepunkt: 196…198 °C (1013 hPa) Schmelzpunkt: –13 °C Dichte: 1,115 g/cm³ (20 °C) Flammpunkt: 116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00002885 CAS-Nummer: 107-21-1 EINECS: 203-473-3 REACH: 01-2119456816-28 Merck Index: 13,03832 |
|
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use | Passes Test |
Appearance | Passes Test |
Assay (HOCH₂CH₂OH) (by GC) | ≥99 % |
Assay (HOCH₂CH₂OH)(wet analysis) | ≥99 % |
Color (APHA) | ≤10 |
Specific Gravity at 20°/20°C | 1.114 - 1.116 |
Residue after Ignition | ≤0.005 % |
Acidity (as CH₃COOH) | ≤0.01 % |
Water (H₂O)(by Karl Fischer titrn) | ≤0.2 % |
Chloride (Cl) | ≤5 ppm |
Trace Impurities - Iron (Fe) | ≤0.2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...