Ethylenglycol ≥99,0% (durch GC), BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Glycol, 1,2-Dihydroxyethan, 1,2-Ethandiol

9300-03EA 320.46 EUR
9300-03 9300-01 9300-07 9300-R
Ethylenglycol ≥99,0% (durch GC), BAKER ANALYZED®, J.T.Baker®
Ethylenglycol
Formel: HOCH₂CH₂OH
Siedepunkt: 196…198 °C (1013 hPa)
Schmelzpunkt: –13 °C
Dichte: 1,115 g/cm³ (20 °C)
Flammpunkt: 116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002885
CAS-Nummer: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Assay (HOCH₂CH₂OH) (by GC) ≥99 %
Assay (HOCH₂CH₂OH)(wet analysis) ≥99 %
Color (APHA) ≤10
Specific Gravity at 20°/20°C 1.114 - 1.116
Residue after Ignition ≤0.005 %
Acidity (as CH₃COOH) ≤0.01 %
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR