Tris-EDTA (TE) buffer solution for molecular biology, pH 7,4, BioUltra, Supelco®
Lieferant: Merck
93302-100ML
93302-100MLEA 40.8 EUR
93302-100ML
Tris-EDTA (TE) buffer solution for molecular biology, pH 7,4, BioUltra, Supelco®
Puffer Biologische Puffer
Tris-EDTA buffer solution has been used for the dilution of DNA samples.[1] It has been used for the elution of purified vector.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7524677cc71a985d71e555814f100d8e6e3b870d&vid=b14f5051142b54a9453694e2696269ce09fb5381&prid=web&clid=SAPDAM