2-Propanol 99,7 GC, corrected for water content, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Dimethylcarbinol, IPA, Isopropylalkohol, 2-Hydroxypropan, Isopropanol
Inert_Gas_filled
9334-03EA
0
EUR
9334-03
2-Propanol 99,7 GC, corrected for water content, ULTRA RESI-ANALYZED® Organic_Residue_Analysis, J.T.Baker®
2-Propanol
Formel:
(CH₃)₂CHOH Molecular Weight: 60.1 g/mol Siedepunkt: 82 Schmelzpunkt: –89 °C Dichte: 0.786 g/cm³ (20 °C) Flammpunkt: 12 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011674 CAS-Nummer: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3 II REACH: 01-2119457558-25 Merck Index: 13,05228 |
|
Spezifikation Testergebnisse
For Laboratory,Research,or Manufacturing Use |
|
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) | ≤5 |
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) | ≤10 |
Assay (CH₃CHOHCH₃) (by GC, corrected for water) | ≥99.7 % |
Color (APHA) | ≤10 |
Residue after Evaporation | ≤1.0 ppm |
Water (by KF, coulometric) | ≤0.1 % |
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...