Supelco® TraceCERT® Organic Reference Standard, Triclosan
Lieferant: Merck
93453-100MG
93453-100MGEA 82.1 EUR
93453-100MG
Supelco® TraceCERT® Organic Reference Standard, Triclosan
Standards Organische Standards
  • GHS signal: GHS07, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P264 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313