(S)-Trolox methyl ether ≥98,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®

Lieferant: Merck

Synonyme: (S)-6-Methoxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

93510-50MG
93510-50MGEA 160 EUR
93510-50MG
(S)-Trolox methyl ether ≥98,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®
(S)-Trolox methyl ether

Chiral derivatizing agent for alcohols; some methyl substituted primary alcohols which are difficult to separate otherwise, can also be separated.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₁₅H₂₀O₄
Molecular Weight: 264,32 g/mol
MDL: MFCD00171453
CAS-Nummer: 135806-59-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbdab054ba6673dc0daedd66e384f585198c60e9&vid=c45ccebef10f5b968ddf2687b4db50e0b868aa1d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR