Ethylenglycol, CMOS für Mikroelektronik, J.T.Baker®

Lieferant: Avantor

Synonyme: Glycol, 1,2-Dihydroxyethan, 1,2-Ethandiol

9356-09EA 0 EUR
9356-09
Ethylenglycol, CMOS für Mikroelektronik, J.T.Baker®
Ethylenglycol
Formel: HOCH₂CH₂OH
Molecular Weight: 62,07 g/mol
MDL: MFCD00002885
CAS-Nummer: 107-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Microelectronic Use
Assay (HOCH₂CH₂OH) (by GC) ≥99.0 %
Color (APHA) ≤10
Acidity (µeq/g) ≤0.8
Acidity (as CH₃COOH)(by wt) ≤0.01 %
Residue after Ignition ≤0.005 %
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤1 ppm
Phosphate (PO₄) ≤2 ppm
Sulfate (SO₄) ≤2 ppm
Arsenic and Antimony (as As) ≤0.100 ppm
Trace Impurities - Aluminum (Al) ≤100.0 ppb
Trace Impurities - Barium (Ba) ≤100.0 ppb
Trace Impurities - Boron (B) ≤50 ppb
Trace Impurities - Calcium (Ca) ≤300 ppb
Trace Impurities - Chromium (Cr) ≤50 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤50 ppb
Trace Impurities - Iron (Fe) ≤200.0 ppb
Trace Impurities - Lead (Pb) ≤200.0 ppb
Trace Impurities - Lithium (Li) ≤100.0 ppb
Trace Impurities - Magnesium (Mg) ≤100.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Tin (Sn) ≤100 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Zinc (Zn) ≤400.0 ppb
Particle Count - 0.5 µm and greater ≤200 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR