Trypanblau 0,4% in wässriger Lösung, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

M-Clarity™ Qualitätsstufe = MQ200

93595-250ML 93595-50ML
93595-250MLEA 88.9 EUR
93595-250ML 93595-50ML
Trypanblau 0,4% in wässriger Lösung, Sigma-Aldrich®
Trypanblau

Trypan blue is a tetrasulfonated anionic, hydrophilic azo dye derived from toluidine. The non-permeable cell membrane dye is a vital dye that selectively stains dead or non-viable cells, that pass through damaged but not through intact cell membranes.


  • it is a routinely used for viability testing for diverse cell types including frozen sperm and aortic muscle cells exposed to antifungal agents
  • employed as a collagen stain in a Van Gieson procedure
  • To stain amyloid, Trypan blue is considered as a blue alternative to Congo red in Puchtler-Bennhold stains
  • used as a fluorescent tracer of cell populations in embryology
  • used as a tumor promoter modulating permeability of lysosomal membranes


Physical form: 0,4% in 0,81% sodium chloride; 0,06% potassium phosphate, dibasic.

Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
Molecular Weight: 960.82 g/mol
Dichte: 1,007 g/ml at 20 °C
Lagertemperatur: Room Temperature
MDL: MFCD00003969
CAS-Nummer: 72-57-1
EINECS: 200-786-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a50f5a57962f84646b27ddad2e42bcb25b8425a&vid=3a50f5a57962f84646b27ddad2e42bcb25b8425a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Blue to very dark blue
Appearance (form) Liquid
Thin layer chromatogr. Corresponds to standard
Density d20/4 0.997 - 1.017
Wavelength (1) (UV) 600 - 610 nm
Extinction (1%/1 cm) (1) (UV)
Solvent (UV) Methanol
Expiry period 30 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR