Dihexylammonium-acetat-Lösung 0.5 M in Wasser, LiChropur™ für die Ionenpaar-Chromatographie, Supelco®

Lieferant: Merck

Synonyme: IPC-DHAA

93645-25ML
93645-25MLEA 128 EUR
93645-25ML
Dihexylammonium-acetat-Lösung 0.5 M in Wasser, LiChropur™ für die Ionenpaar-Chromatographie, Supelco®
Dihexylammonium acetate

Verpackung: Clear borosilicate glass, high hydrolytic resistance (Type I).

Formel: C₁₄H₃₁NO₂
Molecular Weight: 245,41 g/mol
Dichte: 0,992...0.996 g/cm³ (20 °C)
MDL: MFCD03093619
CAS-Nummer: 366793-17-1

Bestellen

Spezifikation Testergebnisse

Appearance (Color) Colorless
Appearance (Form) Liquid
Ttration (Method) Non Aqueous Titration
Titration (Molarity) 0.45 - 0.55 mol/l
Density D20/4 0.992 - 0.996
Refractive Index N20/D 1.350 - 1.354
Test by UV Spectroscopy Corresponds to Requirements
Residue (Filter Test) No Residue
Filtertest (Method) Neat
Recommended Retest Period 42 Months

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR