Salpetersäure 67 - 70% (w/w), BAKER INSTRA-ANALYZED® Plus für die Spurenanalytik von Metallen, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Nitric acid solution

Veränderungsmanagement-Kategorie = N

9368.0500EA 149.74 EUR
9368.0500 9368.1000 9368.2500
Salpetersäure 67 - 70% (w/w), BAKER INSTRA-ANALYZED® Plus für die Spurenanalytik von Metallen, J.T.Baker®
Salpetersäure
Formel: HNO₃
Siedepunkt: 120,5 °C (1013 hPa)
Schmelzpunkt: –42 °C
Dichte: 1,4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 67 - 69 %
Color (APHA) ≤10
Assay (HNO₃)(w/w) 67 - 70 %
Chloride (Cl) ≤0.2 ppm
Total Phosphorus (P) ≤0.01 ppm
Total Sulphur (S) ≤0.3 ppm
Trace Impurities - Aluminum (Al) ≤1 ppb
Trace Impurities - Antimony (Sb) ≤0.5 ppb
Trace Impurities - Arsenic (As) ≤0.5 ppb
Trace Impurities - Barium (Ba) ≤0.1 ppb
Trace Impurities - Beryllium (Be) ≤0.1 ppb
Trace Impurities - Bismuth (Bi) ≤0.1 ppb
Trace Impurities - Cadmium (Cd) ≤0.5 ppb
Trace Impurities - Calcium (Ca) ≤1 ppb
Trace Impurities - Cerium (Ce) ≤0.1 ppb
Trace Impurities - Cesium (Cs) ≤0.1 ppb
Trace Impurities - Cobalt (Co) ≤0.1 ppb
Trace Impurities - Copper (Cu) ≤0.5 ppb
Trace Impurities - Dysprosium (Dy) ≤0.1 ppb
Trace Impurities - Erbium (Er) ≤0.1 ppb
Trace Impurities - Europium (Eu) ≤0.1 ppb
Trace Impurities - Gadolinium (Gd) ≤0.1 ppb
Trace Impurities - Gallium (Ga) ≤0.1 ppb
Trace Impurities - Germanium (Ge) ≤0.1 ppb
Trace Impurities - Gold (Au) ≤0.1 ppb
Trace Impurities - Hafnium (Hf) ≤0.1 ppb
Trace Impurities - Holmium (Ho) ≤0.1 ppb
Trace Impurities - Indium (In) ≤0.1 ppb
Trace Impurities - Iron (Fe) ≤1 ppb
Trace Impurities - Lanthanum (La) ≤0.1 ppb
Trace Impurities - Lead (Pb) ≤0.1 ppb
Trace Impurities - Lithium (Li) ≤0.1 ppb
Trace Impurities - Lutetium (Lu) ≤0.1 ppb
Trace Impurities - Magnesium (Mg) ≤1 ppb
Trace Impurities - Manganese (Mn) ≤0.1 ppb
Trace Impurities - Mercury (Hg) ≤0.1 ppb
Trace Impurities - Molybdenum (Mo) ≤0.1 ppb
Trace Impurities - Neodymium (Nd) ≤0.1 ppb
Trace Impurities - Nickel (Ni) ≤0.5 ppb
Trace Impurities - Niobium (Nb) ≤0.1 ppb
Trace Impurities - Palladium (Pd) ≤0.5 ppb
Trace Impurities - Platinum (Pt) ≤0.5 ppb
Trace Impurities - Potassium (K) ≤1 ppb
Trace Impurities - Praseodymium (Pr) ≤0.1 ppb
Trace Impurities - Rhenium (Re) ≤0.1 ppb
Trace Impurities - Rhodium (Rh) ≤0.5 ppb
Trace Impurities - Rubidium (Rb) ≤0.1 ppb
Trace Impurities - Ruthenium (Ru) ≤0.5 ppb
Trace Impurities - Samarium (Sm) ≤0.1 ppb
Trace Impurities - Scandium (Sc) ≤0.1 ppb
Trace Impurities - Selenium (Se) ≤1 ppb
Trace Impurities - Silver (Ag) ≤0.1 ppb
Trace Impurities - Sodium (Na) ≤1 ppb
Trace Impurities - Strontium (Sr) ≤0.1 ppb
Trace Impurities - Tantalum (Ta) max. 1
Trace Impurities - Tellurium (Te) ≤0.1 ppb
Trace Impurities - Terbium (Tb) ≤0.1 ppb
Trace Impurities - Thallium (Tl) ≤0.1 ppb
Trace Impurities - Thorium (Th) ≤0.1 ppb
Trace Impurities - Thulium (Tm) ≤0.1 ppb
Trace Impurities - Tin (Sn) ≤0.5 ppb
Trace Impurities - Titanium (Ti) ≤0.5 ppb
Trace Impurities - Tungsten (W) ≤0.1 ppb
Trace Impurities - Uranium (U) ≤0.1 ppb
Trace Impurities - Vanadium (V) ≤0.5 ppb
Trace Impurities - Ytterbium (Yb) ≤0.1 ppb
Trace Impurities - Yttrium (Y) ≤0.1 ppb
Trace Impurities - Zinc (Zn) ≤0.5 ppb
Trace Impurities - Zirconium (Zr) ≤0.1 ppb
Ytterbium (Yb) max. 0.1
Zinc (Zn) max. 0.5
Zirconium (Zr) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR