Supelco® TraceCERT® Organic Reference Standard, ʟ-Proline
Lieferant: Merck
93693-100MG
93693-100MGEA
86.4
EUR
93693-100MG
Supelco® TraceCERT® Organic Reference Standard, ʟ-Proline
Standards
Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.