Essigsäure (Eisessig) ≥99%, BAKER INSTRA-ANALYZED® Plus für die Spurenanalytik von Metallen, J.T.Baker®

Lieferant: Avantor
Gefahr

9375.0500EA 148.45 EUR
9375.0500 9375.1000 9375.2500
Essigsäure (Eisessig) ≥99%, BAKER INSTRA-ANALYZED® Plus für die Spurenanalytik von Metallen, J.T.Baker®
Essigsäure

For trace metals analysis, ICP, environmental, pharmaceutical and low metals.

Formel: H₃CCOOH
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,05 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Color (APHA) ≤10
Assay (CH₃COOH, w/w) ≥99 %
Chloride (Cl) ≤1 ppm
Phosphate (PO₄) ≤1 ppm
Sulfate (SO₄) ≤0.5 ppm
Substances Reducing Dichromate (K₂Cr₂O₇) Passes Test
Substances Reducing Permanganate (KMnO₄) Passes Test
Trace Impurities - Aluminum (Al) ≤1 ppb
Trace Impurities - Antimony (Sb) ≤0.5 ppb
Trace Impurities - Arsenic (As) ≤0.5 ppb
Trace Impurities - Barium (Ba) ≤0.5 ppb
Trace Impurities - Beryllium (Be) ≤0.1 ppb
Trace Impurities - Bismuth (Bi) ≤0.1 ppb
Trace Impurities - Cadmium (Cd) ≤0.5 ppb
Trace Impurities - Calcium (Ca) ≤1 ppb
Trace Impurities - Cerium (Ce) ≤0.1 ppb
Trace Impurities - Cesium (Cs) ≤0.1 ppb
Trace Impurities - Chromium (Cr) ≤1 ppb
Trace Impurities - Cobalt (Co) ≤0.1 ppb
Trace Impurities - Copper (Cu) ≤0.5 ppb
Trace Impurities - Dysprosium (Dy) ≤0.1 ppb
Trace Impurities - Erbium (Er) ≤0.1 ppb
Trace Impurities - Europium (Eu) ≤0.1 ppb
Trace Impurities - Gadolinium (Gd) ≤0.1 ppb
Trace Impurities - Gallium (Ga) ≤0.1 ppb
Trace Impurities - Germanium (Ge) ≤0.5 ppb
Trace Impurities - Hafnium (Hf) ≤0.1 ppb
Trace Impurities - Holmium (Ho) ≤0.1 ppb
Trace Impurities - Indium (In) ≤0.1 ppb
Trace Impurities - Iron (Fe) ≤1 ppb
Trace Impurities - Lanthanum (La) ≤0.1 ppb
Trace Impurities - Lead (Pb) ≤0.1 ppb
Trace Impurities - Lithium (Li) ≤0.1 ppb
Trace Impurities - Lutetium (Lu) ≤0.1 ppb
Trace Impurities - Magnesium (Mg) ≤0.5 ppb
Trace Impurities - Manganese (Mn) ≤0.5 ppb
Trace Impurities - Mercury (Hg) ≤1 ppb
Trace Impurities - Molybdenum (Mo) ≤0.5 ppb
Trace Impurities - Neodymium (Nd) ≤0.1 ppb
Trace Impurities - Nickel (Ni) ≤0.5 ppb
Trace Impurities - Platinum (Pt) ≤0.5 ppb
Trace Impurities - Potassium (K) ≤1 ppb
Trace Impurities - Praseodymium (Pr) ≤0.1 ppb
Trace Impurities - Rhenium (Re) ≤0.1 ppb
Trace Impurities - Rhodium (Rh) ≤0.5 ppb
Trace Impurities - Rubidium (Rb) ≤0.1 ppb
Trace Impurities - Ruthenium (Ru) ≤0.5 ppb
Trace Impurities - Samarium (Sm) ≤0.1 ppb
Trace Impurities - Scandium (Sc) ≤0.1 ppb
Trace Impurities - Selenium (Se) ≤1 ppb
Trace Impurities - Silver (Ag) ≤1 ppb
Trace Impurities - Sodium (Na) ≤1 ppb
Trace Impurities - Strontium (Sr) ≤0.5 ppb
Trace Impurities - Tellurium (Te) ≤0.5 ppb
Trace Impurities - Terbium (Tb) ≤0.1 ppb
Trace Impurities - Thallium (Tl) ≤0.1 ppb
Trace Impurities - Thorium (Th) ≤0.1 ppb
Trace Impurities - Thulium (Tm) ≤0.1 ppb
Trace Impurities - Tin (Sn) ≤0.5 ppb
Trace Impurities - Titanium (Ti) ≤0.5 ppb
Trace Impurities - Tungsten (W) ≤0.5 ppb
Trace Impurities - Uranium (U) ≤0.1 ppb
Trace Impurities - Vanadium (V) ≤0.5 ppb
Trace Impurities - Ytterbium (Yb) ≤0.1 ppb
Trace Impurities - Yttrium (Y) ≤0.1 ppb
Trace Impurities - Zinc (Zn) ≤1 ppb
Trace Impurities - Zirconium (Zr) ≤0.1 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR