Wasser, BAKER INSTRA-ANALYZED® Plus, J.T.Baker®

Lieferant: Avantor

9381.1000EA 206.41 EUR
9381.1000
Wasser, BAKER INSTRA-ANALYZED® Plus, J.T.Baker®
Wasser
Formel: H₂O
MDL: MFCD00011332
CAS-Nummer: 7732-18-5

Bestellen

Spezifikation Testergebnisse

Colour (Hazen) max. 5
LC Gradient Suitability Test passes test
Optical Absorbance
at 200 nm max. 0.01
at 254 nm max. 0.005
at 300 nm max. 0.005
at 400 nm max. 0.005
Trace Impurities
Fluorescence (as Quinine) max. 100 ppt
Residue after Evaporation max. 1 ppm
Resistivity (at time of bottling) min. 18 megaohm
Total Organic Carbon max. 20 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR