Flußsäure 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus für die Spurenanalytik von Metallen, J.T.Baker®

Lieferant: Avantor
Gefahr


Veränderungsmanagement-Kategorie = N

9387.0500EA 306.37 EUR
9387.0500
Flußsäure 47 - 51% (w/w), BAKER INSTRA-ANALYZED® Plus für die Spurenanalytik von Metallen, J.T.Baker®
Hydrofluoric acid
Formel: HF
Molecular Weight: 20,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00011346
CAS-Nummer: 7664-39-3
EINECS: 231-634-8
UN: 1790
ADR: 8,I

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 47 - 51 %
Color (APHA) ≤10
Assay (HF)(w/w) 47 - 51 %
Chloride (Cl) ≤4 ppm
Fluosilicic Acid (SiF₆) ≤20 ppm
Total Phosphorus (P) ≤0.05 ppm
Total Sulphur (S) ≤0.1 ppm
Trace Impurities - Aluminum (Al) ≤1 ppb
Trace Impurities - Antimony (Sb) ≤0.2 ppb
Trace Impurities - Arsenic (As) ≤0.5 ppb
Trace Impurities - Barium (Ba) ≤0.1 ppb
Trace Impurities - Beryllium (Be) ≤0.1 ppb
Trace Impurities - Bismuth (Bi) ≤0.1 ppb
Trace Impurities - Boron (B) ≤1 ppb
Trace Impurities - Cadmium (Cd) ≤0.1 ppb
Trace Impurities - Calcium (Ca) ≤1 ppb
Trace Impurities - Cerium (Ce) ≤0.1 ppb
Trace Impurities - Cesium (Cs) ≤0.1 ppb
Trace Impurities - Chromium (Cr) ≤1 ppb
Trace Impurities - Cobalt (Co) ≤0.1 ppb
Trace Impurities - Copper (Cu) ≤0.5 ppb
Trace Impurities - Dysprosium (Dy) ≤0.1 ppb
Trace Impurities - Erbium (Er) ≤0.1 ppb
Trace Impurities - Europium (Eu) ≤0.1 ppb
Trace Impurities - Gadolinium (Gd) ≤0.1 ppb
Trace Impurities - Gallium (Ga) ≤0.1 ppb
Trace Impurities - Germanium (Ge) ≤0.1 ppb
Trace Impurities - Gold (Au) ≤0.2 ppb
Trace Impurities - Hafnium (Hf) ≤0.1 ppb
Trace Impurities - Holmium (Ho) ≤0.1 ppb
Trace Impurities - Indium (In) ≤0.1 ppb
Trace Impurities - Iron (Fe) ≤1 ppb
Trace Impurities - Lanthanum (La) ≤0.1 ppb
Trace Impurities - Lead (Pb) ≤0.1 ppb
Trace Impurities - Lithium (Li) ≤0.1 ppb
Trace Impurities - Lutetium (Lu) ≤0.1 ppb
Trace Impurities - Magnesium (Mg) ≤1 ppb
Trace Impurities - Manganese (Mn) ≤0.1 ppb
Trace Impurities - Mercury (Hg) ≤1 ppb
Trace Impurities - Molybdenum (Mo) ≤0.1 ppb
Trace Impurities - Neodymium (Nd) ≤0.1 ppb
Trace Impurities - Nickel (Ni) ≤0.5 ppb
Trace Impurities - Niobium (Nb) ≤0.1 ppb
Trace Impurities - Palladium (Pd) max. 0.1
Trace Impurities - Platinum (Pt) max. 1
Trace Impurities - Potassium (K) ≤1 ppb
Trace Impurities - Praseodymium (Pr) ≤0.1 ppb
Trace Impurities - Rhenium (Re) ≤0.1 ppb
Trace Impurities - Rhodium (Rh) ≤0.1 ppb
Trace Impurities - Rubidium (Rb) ≤0.1 ppb
Trace Impurities - Ruthenium (Ru) ≤0.1 ppb
Trace Impurities - Samarium (Sm) ≤0.1 ppb
Trace Impurities - Scandium (Sc) ≤0.1 ppb
Trace Impurities - Selenium (Se) ≤1 ppb
Trace Impurities - Silver (Ag) ≤0.5 ppb
Trace Impurities - Sodium (Na) ≤1 ppb
Trace Impurities - Strontium (Sr) ≤0.1 ppb
Trace Impurities - Tantalum (Ta) max. 0.1
Trace Impurities - Tellurium (Te) ≤0.1 ppb
Trace Impurities - Terbium (Tb) ≤0.1 ppb
Trace Impurities - Thallium (Tl) ≤0.1 ppb
Trace Impurities - Thorium (Th) ≤0.1 ppb
Trace Impurities - Thulium (Tm) ≤0.1 ppb
Trace Impurities - Tin (Sn) ≤0.5 ppb
Trace Impurities - Titanium (Ti) ≤1 ppb
Trace Impurities - Tungsten (W) ≤0.5 ppb
Trace Impurities - Uranium (U) ≤0.1 ppb
Trace Impurities - Vanadium (V) ≤0.1 ppb
Trace Impurities - Ytterbium (Yb) ≤0.1 ppb
Trace Impurities - Yttrium (Y) ≤0.1 ppb
Trace Impurities - Zinc (Zn) ≤1 ppb
Trace Impurities - Zirconium (Zr) ≤0.1 ppb
Samarium (Sm) max. 0.1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR