Supelco® TraceCERT® Organic Reference Standard, Chrysene
Lieferant: Merck
94035-50MG
94035-50MGEA 81.8 EUR
94035-50MG
Supelco® TraceCERT® Organic Reference Standard, Chrysene
Standards Organische Standards
  • GHS signal: GHS08, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P201 - P273 - P308 + P313
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cdcc34e7b57fef8eb9f16c97d13a4e6d7f05b3e6&vid=cdcc34e7b57fef8eb9f16c97d13a4e6d7f05b3e6&prid=web&clid=SAPDAM