Supelco® TraceCERT® Organic Reference Standard, Chlorpyrifos
Lieferant: Merck
94114-100MG
94114-100MGEA
76.7
EUR
94114-100MG
Supelco® TraceCERT® Organic Reference Standard, Chlorpyrifos
Standards
Organische Standards
- GHS signal: GHS06, GHS09
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P264 - P270 - P273 - P301 + P310 - P391 - P405