Lewatit® VP OC 1065 ~0,5 mm (schwach basischer Anionenaustauscher), Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
94136-100G-F
94136-100G-FEA
47.5
EUR
94136-100G-F
Lewatit® VP OC 1065 ~0,5 mm (schwach basischer Anionenaustauscher), Supelco®
Lewatit® VP OC 1065
Adsorber resin functionalised with benzyl amine groups.
- Loss: ~50% loss on drying
- Matrix: Styrene-divinylbenzene
- Matrix active group: Trimethylamine
- Particle size: ~0,5 mm
- Capacity: 1,4 meq/ml