Alizarinrot S (Natriumsalz)
Lieferant: VWR Chemicals
|
Achtung
|
9436-25GEA
132
EUR
9436-25G
Alizarinrot S (Natriumsalz)
Alizarinrot S (Natriumsalz)
Formel:
C₁₄H₇NaO₇S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013049 CAS-Nummer: 130-22-3 EINECS: 204-981-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...