Supelco® TraceCERT® Organic Reference Standard, Indeno[1,2,3-cd]pyrene
Lieferant: Merck
94377-10MG
94377-10MGEA 89.9 EUR
94377-10MG
Supelco® TraceCERT® Organic Reference Standard, Indeno[1,2,3-cd]pyrene
Standards Organische Standards
  • GHS signal: GHS08
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P201 - P202 - P280 - P308 + P313 - P405 - P501
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=668409468299af0996e63090117fbeca8f471e35&vid=668409468299af0996e63090117fbeca8f471e35&prid=web&clid=SAPDAM