Kristallviolett solution_SC Grams Microscopy, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: Hexamethyl-p-rosaniliniumchlorid, Hexamethylen-p-rosanilinchlorid, blaues Pyoktanin, Basic Violet 3, Hexamethylviolett, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammoniumchlorid, Methylviolett 10B, Methylrosanilin, Methylrosanilium, 4,4',4''-Tri(dimethylamino)trityliumchlorid, CI 42555, Methylrosaniliumchlorid, Gentianaviolett

MQ200

94448-2.5L-F 94448-250ML-F
94448-2.5L-FEA 90.8 EUR
94448-2.5L-F 94448-250ML-F
Kristallviolett solution_SC Grams Microscopy, Sigma-Aldrich®
Kristallviolett

Gram’s crystal violet solution, also known as gentian violet or hexamethyl pararosaniline, is a basic, cationic, acidotropic aminoarylmethane dye. It is a hexa-N-methylated triaminotriphenylmethane with a moderately sized lipophilic cation and a slightly nonplanar conjugated system. It is used in bacteriology as the primary stain in Gram staining to differentiate Gram-positive and Gram-negative bacteria. It is known to possess antibacterial and antifungal properties.


  • Crystal violet is routinely used for Gram staining and its variants, staining amyloid, bacterial components, and vascular plant tissues
  • It is used in polychrome staining of epoxy resin sections, viability staining of cultured neurons, and confocal optical sectioning to analyse meiotic structures
  • It is also employed in the acridine orange-crystal violet staining of intracellular bacteria, microsporidian spores, and cytological smears


The antimicrobial effects of crystal violet are caused by free radical generation and the formation of unionised dye-bacteria complexes. It also inhibits bacterial protein synthesis and cell wall formation and is especially effective against Gram-positive bacteria.

Formel: C₂₅H₃₀ClN₃
Molecular Weight: 407.99 g/mol
Dichte: 0,981 g/ml (20 °C)
Flammpunkt: 34 °C
Lagertemperatur: Room Temperature
MDL: MFCD00011750
CAS-Nummer: 548-62-9
UN: 1170
ADR: 3 II
Merck Index: 13,04406

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Purple to very dark purple
Appearance (form) Liquid
Thin layer chromatogr. Corresponds to standard
Density D20/4 0.970 - 0.985
Wavelength (1) (UV) 585 - 595 nm
Extinction (1%/1 cm) (1) (UV) Report result
Solvent (UV) Water
Expiry period 36 months


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR