Gram′s safranin solution zur Gramfärbung, für die Mikroskopie, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
M-Clarity™ Qualitätsstufe = MQ100
Gram’s safranin solution, also known as Safranin T or Basic Red 2, is a ready-to-use staining solution that comprises 10% safranin in ethanol. Safranin is a basic, cationic, azine dye containing a planar, diaminophenazine chromophore, with a phenyl substituent attached to a ring nitrogen. The trimethyl cationic form of safranine is lipophilic, whereas the dimethyl forms are weakly hydrophilic. It is most notably used in bacteriology as a counterstain in Gram staining to differentiate between Gram-positive and Gram-negative bacteria.
- Ready-to-use solution
- Modified and designed in such a way that staining can be carried out in staining cells, on the staining rack, and in automated staining systems
Gram’s safranin solution has been used in the following applications:
as a non-toxic replacement for crystal violet for the quantification of biofilm formation
to develop a hyaluronic acid visualisation method
to study improved β-lactam susceptibility against biofilm-embedded staphylococcus aureus by 2-aminothiazole
Formel:
C₂₀H₁₉ClN₄ Dichte: 0,986 g/ml at 20 °C Flammpunkt: 46 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00011759 CAS-Nummer: 477-73-6 |
|
Spezifikation Testergebnisse
Appearance (colour) | Red to very dark red |
Appearance (form) | Liquid |
Thin layer chromatogr. | Corresponds to standard |
Density D20/4 | 0.984 - 0.988 |
Refractive index N20/D | 1.338 - 1.340 |
Wavelength (1) (UV) | 525 - 535 nm |
Extinction (1%/1 cm) (1) (UV) | Report result |
Solvent (UV) | Ethanol 50% |
Expiry period | 36 months |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...