Organic reference standard, Vanadate-molybdate reagent, for phosphate determination, p.a., Supelco®
Lieferant: Merck
94685-500ML
94685-500MLEA
54.4
EUR
94685-500ML
Organic reference standard, Vanadate-molybdate reagent, for phosphate determination, p.a., Supelco®
Standards
Organische Standards
Vanadate-molybdate reagent can be prepared by dissolving ammonium molybdate and ammonium vanadate in distilled water along with concentrated nitric acid.
Vanadate-molybdate reagent has been used to determine phosphorous using atomic absorption spectrometer, by vanadium phosphomolybdate method from cashew apple juice samples.
Reagent, suitable for eliminating interfering effects by phosphate and silicate in the AAS determination of Ca and Mg.