Melting point standards, methyltriphenylphosphoniumbromide, traceable to primary standards, Supelco®
Lieferant: Merck
94993-5G
94993-5GEA
51.5
EUR
94993-5G
Melting point standards, methyltriphenylphosphoniumbromide, traceable to primary standards, Supelco®
Standards
Schmelzpunkt-Standards
Mettler-Toledo Calibration substance ME 30130598, Methyltriphenylphosphoniumbromide, tracable to primary standards (LGC).
Suitable for dropping point calibration.