Melting point standards, methyltriphenylphosphoniumbromide, traceable to primary standards, Supelco®
Lieferant: Merck
94993-5G
94993-5GEA 51.5 EUR
94993-5G
Melting point standards, methyltriphenylphosphoniumbromide, traceable to primary standards, Supelco®
Standards Schmelzpunkt-Standards
Mettler-Toledo Calibration substance ME 30130598, Methyltriphenylphosphoniumbromide, tracable to primary standards (LGC).
Suitable for dropping point calibration.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4897cf256c13cf627d692d2ae610ac03ea37df00&vid=4897cf256c13cf627d692d2ae610ac03ea37df00&prid=web&clid=SAPDAM