Apiezon® Wachs W, Stangen
Lieferant: M I MATERIALS
Vacuum sealing and mounting wax to produce rigid permanent or semi-permanent sealed joints.
- Good resistance to water and attack by acids and alkalis
- Low vapour pressure (4.5 x 10-9 torr at 20°C)
- High melting point (softens between 80-90°C)
Bestellinformation: Wie Apiezon Wachs-W-Sticks (oben), jedoch als 1 kg Block anstatt in Stick-Form