Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Lieferant: Honeywell Chemicals
95164-250ML
95164-250MLEA
357.87
EUR
95164-250ML
Cesium chloride - lanthanum chloride buffer, for flame AAS, TraceSELECT™, Fluka™
Puffer
Ionisationspuffer
TraceSELECT™, for flame atomic absorption spectrometry.
- Packaged in high-quality containers appropriate for each product
- The reagents are produced and bottled under clean-room conditionsions
Lanthanum chloride (LaCl3) 99,0 - 101,0 g/l
Cesium chloride (Cscl) 99,0 - 101,0 g/l
Density (D 20/4) 1,152 - 1,176
Calcium (Ca) max. 0,5 ppm