Essigsäure (Eisessig) ≥ 90% 99,5-100,5% USP, FCC, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Ethansäure

9522-02EA 101.72 EUR
9522-02 9522-05 9522-33 9522-07 9522-03 9522-15 9522-08
Essigsäure (Eisessig) ≥ 90% 99,5-100,5% USP, FCC, J.T.Baker®
Essigsäure
Formel: H₃CCOOH
Molecular Weight: 60.05 g/mol
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1.05 g/cm³ (20 °C)
Flammpunkt: 38.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8 II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product Passes Test
Meets U.S.P Requirements = 15.6 °C
Meets F.C.C. Requirements = 1.0
GMP Manufactured Product Passes Test
IMPORTANT: Material will freeze if stored below 17 °C (63°F). Passes Test
CAUTION: For Manufacturing, processing or repackaging = 5 ppm
Bulk Pharmaceutical Chemical Passes Test
Bulk Food Chemical 99.5 - 100.5 %
USP - Identification Passes Test
USP - Congealing Temperature (solidification point) ≥15.6 °C
USP - Nonvolatile Residue (USP), mg/20 mL ≤1.0
USP - Chloride (Cl) Passes Test
USP - Sulfate (SO₄) Passes Test
USP - Readily Oxidizable Substances Passes Test
USP - Assay (CH₃COOH) 99.5 - 100.5 %
FCC - Identification Passes Test
FCC - Congealing Temperature (solidification point) ≥15.6 °C
FCC - Nonvolatile Residue (FCC) ≤0.005 %
FCC - Readily Oxidizable Substances Passes Test
FCC - Assay (CH₃COOH) 99.5 - 100.5 %
FCC - Lead (Pb) ≤0.5 mg/kg
Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or
produced by the production process.
Preserve in tight containers, and store at room temperature
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR