Essigsäure (Eisessig) 99,5-100,05% USP, Multikompendial, J.T.Baker®

Lieferant: Avantor
Gefahr

9526-01EA 109.06 EUR
9526-01 9526-07 9526-20 9526-03 9526-33 9526-26
Essigsäure (Eisessig) 99,5-100,05% USP, Multikompendial, J.T.Baker®
Essigsäure
Formel: H₃CCOOH
Siedepunkt: 118 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,05 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements
GMP Manufactured Product
Meets J.P. Chemical Specifications
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical = 15.6 °C
USP - Identification Passes Test
USP - Congealing Temperature ≥15.6 °C
USP - Nonvolatile Residue (USP), mg/20 mL ≤1.0
USP - Chloride (Cl) Passes Test
USP - Sulfate (SO₄) Passes Test
USP - Readily Oxidizable Substances Passes Test
USP - Assay (CH₃COOH) 99.5 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance Passes Test
EP/BP - Freezing Point ≥14.8 °C
EP/BP - Iron (Fe) ≤5 ppm
EP/BP - Chloride (Cl) ≤25 ppm
EP/BP - Sulfate (SO₄) ≤50 ppm
EP/BP - Reducing Substances Passes Test
EP/BP - Residue on Evaporation ≤0.01 %
EP/BP - Assay (CH₃COOH) 99.0 - 100.5 %
JP - Assay (CH₃COOH) 99.0 - 101.0 %
JP - Identification Passes Test
JP - Congealing Point ≥14.5 °C
JP - Nonvolatile Residue, mg/10 mL ≤1.0
JP - Chloride (Cl) Passes Test
JP - Sulfate (SO₄) Passes Test
JP - Heavy Metals (as Pb) ≤10 ppm
JP - Reducing Substances Passes Test
Residual Solvents: No Class 1, 2 or 3 residual solvents are used in or
produced by the production process.
Preserve in tight containers, and store at room temperature
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR