Wasser for HPCE, for luminescence, suitable for UV/Vis spectroscopy, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

95283-100ML 95283-1L
95283-100MLEA 32.8 EUR
95283-100ML 95283-1L
Wasser for HPCE, for luminescence, suitable for UV/Vis spectroscopy, Supelco®
Wasser
Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3fc5b8c508b689bd300e5109ec8084e515240d43&vid=60245314e6bb0a0e76427301c61dd541f6fbd765&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1eb00964e70724f101d4f378d2f4b23d5af316d9&vid=24cd6362eb60659f9c641266bdba6c8f1240ff74&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR